Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Japanese Encephalitis Envelope Protein Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111964
Description
Japanese Encephalitis Envelope Protein Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Japanese encephalitis (JE) is an infection of the brain caused by the Japanese encephalitis virus (JEV). While most infections result in little or no symptoms, occasional inflammation of the brain occurs. In these cases, symptoms may include headache, vomiting, fever, confusion and seizures. The JEV envelope protein (E) facilitates cellular attachment and membrane fusion and is the primary target of neutralizing antibodies.
Specifications
| Japanese Encephalitis Envelope Protein | |
| Polyclonal | |
| Unconjugated | |
| Japanese Encephalitis | |
| Recombinant protein encompassing a sequence within the center region of Envelope protein (JEV) (Japanese Encephalitis Virus strain Jaoars982). The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.18 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction