Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Japanese Encephalitis virus Capsid Protein Antibody (GT1178), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP313585
This item is not returnable.
View return policy
Description
Japanese Encephalitis virus Capsid Protein Monoclonal antibody specifically detects Japanese Encephalitis virus Capsid Protein in Virus samples. It is validated for Western BlotSpecifications
| Japanese Encephalitis virus Capsid Protein | |
| Monoclonal | |
| Unconjugated | |
| Capsid protein C, Core protein, JEV Capsid Protein | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of Core protein C (JEV). (Japanese Encephalitis Virus strain Jaoars982) The exact sequence is proprietary. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot | |
| GT1178 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction