Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Japanese Encephalitis virus Capsid Protein Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP312875
This item is not returnable.
View return policy
Description
Japanese Encephalitis virus Capsid Protein Polyclonal antibody specifically detects Japanese Encephalitis virus Capsid Protein in Virus samples. It is validated for Western BlotSpecifications
| Japanese Encephalitis virus Capsid Protein | |
| Polyclonal | |
| PBS, 20% Glycerol (pH7) | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot | |
| Unconjugated | |
| Capsid protein C, Core protein, JEV Capsid Protein | |
| Full length Japanese Encephalitis virus Capsid Protein Recombinant protein (Japanese Encephalitis Virus strain Jaoars982) | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction