Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Japanese encephalitis virus Mouse anti-Virus, Unconjugated, Clone: HM467, Abnova™
Mouse Monoclonal Antibody.
Supplier: Abnova Corporation MAB21914
Description
Japanese encephalitis virus Monoclonal antibody specifically detects Japanese encephalitis virus in Viruses samples. It is validated for ELISA, LFIA.Specifications
Japanese encephalitis virus | |
Monoclonal | |
Unconjugated | |
Purified Japanese encephalitis virus | |
Primary | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
IgG1 κ |
ELISA, Lateral Flow | |
HM467 | |
Mouse | |
1 mg | |
Virus | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction