Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Japanese Encephalitis Virus NS1 Monoclonal Antibody (3594)
GREENER_CHOICE

Catalog No. PIMA533269
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533269 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533269 Supplier Invitrogen™ Supplier No. MA533269
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Non-reactive with: TBE virus, Yellow Fever virus, West Nile virus, Dengue viruses (1-4), Zika virus and Chikungunya virus. ELISA pair: MA5-33268 (capture) and MA5-33269 (conjugate).

Japanese encephalitis virus (JEV) is a flavivirus related to dengue, yellow fever and West Nile viruses, and is spread by mosquitoes. JEV is the main cause of viral encephalitis in many countries of Asia with an estimated 68000 clinical cases every year.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Japanese Encephalitis Virus NS1
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 3594
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Japanese Encephalitis
Host Species Mouse
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.