Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Japanese Encephalitis Virus NS2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532237
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532237 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532237 Supplier Invitrogen™ Supplier No. PA532237
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Japanese encephalitis virus infect BHK-21. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Japanese encephalitis virus (JEV) is mosquito born flavivirus that induces acute encephalitis in tropical and subtropical world.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Japanese Encephalitis Virus NS2B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene Alias Flavivirus; JEV; JEV NS2B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of NS2B (JEV) (Japanese Encephalitis Virus strain Jaoars982). The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.