Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Japanese Encephalitis virus NS4A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP313058
This item is not returnable.
View return policy
Description
Japanese Encephalitis virus NS4A Polyclonal antibody specifically detects Japanese Encephalitis virus NS4A in Virus samples. It is validated for Western BlotSpecifications
| Japanese Encephalitis virus NS4A | |
| Polyclonal | |
| PBS, 20% Glycerol (pH7) | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot | |
| Unconjugated | |
| JEV NS4A, Non-structural protein 4A, NS4A | |
| Recombinant protein encompassing a sequence within the C-terminus region of NS4A (JEV). (Japanese Encephalitis Virus strain Jaoars982) The exact sequence is proprietary. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction