Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Japanese Encephalitis Virus NS4B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532211
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532211 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532211 Supplier Invitrogen™ Supplier No. PA532211
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: BHK21 infected JEV. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Japanese encephalitis virus (JEV) is mosquito born flavivirus that induces acute encephalitis in tropical and subtropical world.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Japanese Encephalitis Virus NS4B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.76 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Alias Flavivirus; JEV; NS4B
Host Species Rabbit
Immunogen Full length Japanese encephalitis virus JEV NS4b Recombinant protein (Japanese Encephalitis Virus strain Jaoars982).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.