Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Jarid1A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 5 publications
Supplier: Novus Biologicals NB11040499
Description
Jarid1A Polyclonal specifically detects Jarid1A in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Jarid1A | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
EC 1.14.11, EC 1.14.11.-, Histone demethylase JARID1A, JARID1AJumonji, AT rich interactive domain 1A (RBP2-like), Jumonji, AT rich interactive domain 1A (RBBP2-like), Jumonji/ARID domain-containing protein 1A, KDM5A, lysine (K)-specific demethylase 5A, RBBP-2, RBBP2lysine-specific demethylase 5A, RBP2retinoblastoma binding protein 2, Retinoblastoma-binding protein 2jumonji, AT rich interactive domain 1A | |
Rabbit | |
192 kDa | |
0.1 mL | |
Primary | |
Based on 100% sequence identity, this antibody is predicted to react with Small-eared galago and Northern white-cheeked gibbon. | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 4-10 ug/mg lysate | |
P29375 | |
KDM5A | |
The immunogen recognized by this antibody maps to a region between residue 1640 and 1690 of human JARID1A/RBP2 using the numbering given in entry NP_005047.1 (GeneID 5927). | |
Affinity Purified | |
RUO | |
5927 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction