Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JARID1A/RBP2 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501572163
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-2163 100 μL
50-157-2164 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-2163 Supplier Bethyl Laboratories, Inc Supplier No. IHC00479
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Dog, Horse, Rabbit, Guinea pig_10141, Panda, Orangutan, Gorilla and Northern w

This gene encodes a member of the Jumonji, AT-rich interactive domain 1 (JARID1) histone demethylase protein family. The encoded protein plays a role in gene regulation through the histone code by specifically demethylating lysine 4 of histone H3. The encoded protein interacts with many other proteins, including retinoblastoma protein, and is implicated in the transcriptional regulation of Hox genes and cytokines. This gene may play a role in tumor progression.

Specifications

Antigen JARID1A/RBP2
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene KDM5A
Gene Accession No. P29375
Gene Alias histone demethylase JARID1A, JARID1A, jumonji, AT rich interactive domain 1A, Jumonji, AT rich interactive domain 1A (RBBP2-like), Jumonji, AT rich interactive domain 1A (RBP2-like), jumonji/ARID domain-containing protein 1A, lysine (K)-specific demethylase 5A, RBBP2, RBBP-2, RBP2, retinoblastoma binding protein 2, retinoblastoma-binding protein 2
Gene Symbols KDM5A
Host Species Rabbit
Immunogen Between 1640 and 1690
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5927
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.