Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JARID1B Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501571980
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1980 10 μL
50-157-1979 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-1980 Supplier Bethyl Laboratories, Inc Supplier No. IHC00370T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Dog, Panda, Orangutan, Monkey, Gorilla, Chimpanzee, Northern white-cheeked gibbon and Smal

KDM5B is a histone demethylase that demethylates ′Lys-4′ of histone H3, thereby playing a central role in histone code. It does not demethylate histone H3 ′Lys-9′ or H3 ′Lys-27′. KDM5B demethylates trimethylated, dimethylated and monomethylated H3 ′Lys-4′. It acts as a transcriptional corepressor for FOXG1B and PAX9. It favors the proliferation of breast cancer cells by repressing tumor suppressor genes such as BRCA1 and HOXA5. In contrast, KDM5B may act as a tumor suppressor for melanoma.

Specifications

Antigen JARID1B
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene KDM5B
Gene Accession No. Q9UGL1
Gene Alias RBBP2H1A, PLU1, PLU-1, CT31, lysine K-specific demethylase 5B, JARID1B, PUT1
Gene Symbols KDM5B
Host Species Rabbit
Immunogen Between 1494 and 1544
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10765
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.