Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JARID2 Monoclonal Antibody (7H1B3)
JARID2 Monoclonal Antibody (7H1B3)
Supplier: Thermo Scientific MA538546
Description
JARID2 Monoclonal Antibody for Western Blot, ICC/IF, Flow, ELISA
Specifications
| JARID2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q92833 | |
| JARID2 | |
| Purified recombinant fragment of human JARID2 (aa: 1097-1246) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
| 7H1B3 | |
| Unconjugated | |
| JARID2 | |
| JARD2 antibody; Jarid2; Jmj; jumonji; jumonji and AT-rich interaction domain containing 2; jumonji homolog; jumonji, AT rich interactive domain 2; jumonji, AT rich interactive domain 2 protein; jumonji/ARID domain-containing protein 2; jumonji-like protein; LOW QUALITY PROTEIN: protein Jumonji; protein Jumonji | |
| Mouse | |
| Protein G | |
| RUO | |
| 3720 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction