Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAZ Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501567470
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-7470 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-7470 Supplier Bethyl Laboratories, Inc Supplier No. A304341A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan

ZNF346, also known as JAZ (just another zinc-finger protein), is a nucleolar, zinc finger protein which preferentially binds to double-stranded (ds) RNA or RNA/DNA hybrids with high affinity via C2H2 zinc fingers. ZNF346 contains four C(2)H(2)-type zinc finger motifs that are connected by long (28-38) amino acid linker sequences. ZNF346 is expressed in all tissues tested and localizes to the nucleus, primarily the nucleolus. ZNF346 is exported by exportin-5 but translocates back into nuclei by a facilitated diffusion mechanism. ZNF346 interacts with ILF3 in an RNA-independent manner and may be involved in cell growth and survival.

Specifications

Antigen JAZ
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene ZNF346
Gene Accession No. Q9UL40
Gene Alias JAZ, Zfp346; double-stranded RNA-binding zinc finger protein JAZ; just another zinc finger protein
Gene Symbols ZNF346
Host Species Rabbit
Immunogen Between 244 and 294
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23567
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.