Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ JAZ11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119952
This item is not returnable.
View return policy
Description
Positive Control: Rice tissue. Subcellular Location: Nucleus.
Repressor of jasmonate responses.
Specifications
JAZ11 | |
Polyclonal | |
Unconjugated | |
LOC4331833 | |
LOC4331833 | |
Synthetic peptide within rice JAZ11 aa 160-209/209. | |
100 μL | |
Primary | |
Plant | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
0 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
4331833 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction