Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JAZF1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA541686

Catalog No. PIPA541686


Only null left
Add to Cart

Description

Description

Peptide sequence: KKRYKNVNGI KYHAKNGHRT QIRVRKPFKC RCGKSYKTAQ GLRHHTINFH Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

JAZF1 is a nuclear protein with three C2H2-type zinc fingers, and functions as a transcriptional repressor. Chromosomal aberrations involving this gene are associated with endometrial stromal tumors.This gene encodes a nuclear protein with three C2H2-type zinc fingers, and functions as a transcriptional repressor. Chromosomal aberrations involving this gene are associated with endometrial stromal tumors. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

JAZF1
Polyclonal
Unconjugated
JAZF1
AI591476; C820002C15; DKFZp761K2222; Jaz1; JAZF zinc finger 1; Jazf1; juxtaposed with another zinc finger gene 1; juxtaposed with another zinc finger protein 1; TAK1-interacting protein 27; TIP27; Zinc finger protein 802; ZNF802
Rabbit
Affinity Chromatography
RUO
221895
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry, Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q86VZ6
JAZF1
Synthetic peptide directed towards the C-terminal of human JAZF1 (aa 181-230).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.