Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ JC-1 Mitochondrial Membrane Potential Dye

Catalog No. 5016951
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-169-51 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-169-51 Supplier Invitrogen™ Supplier No. 65085138
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Used to determine mitochondrial membrane potential in flow cytometry and fluorescent microscopy. Dye selectively enters mitochondria where it reversibly changes color as membrane potentials increase. Property due to the reversible formation of JC-1 aggregates upon membrane polarization.

  • Color changes as membrane potentials increase above values of about 80 to 100 mV
  • Emitted light shifts from 530nm (JC-1 monomeric form) to 590nm (J-aggregate)
  • Each color can be detected using filters for FITC and PE/phycoerythrin/rhodamine, respectively
  • Can be used to indicate the initiation of apoptosis
  • Molecular Weight: 652.23g/mol
  • Peak Excitation: 515nm
  • Peak Emission: 529 and 590nm
  • Should be reconstituted in high quality freshly opened DMSO at 1mg/ml
  • Once reconstituted, it should be stored at −20°C; avoid freeze-thawing
TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Product Type Dye
Product Line eBioscience
Quantity 5 mg
Sub Cellular Localization Mitochondria
Emission Wavelength 515/529&590
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.