Learn More
Invitrogen™ JDP2 Polyclonal Antibody

Description
JDP2 Polyclonal Antibody detects endogenous levels of JDP2 protein.
Specifications
Specifications
| Antigen | JDP2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | JDP2 |
| Gene Accession No. | P97875, Q78E65, Q8WYK2 |
| Gene Alias | c-Jun dimerization protein 2; Jdp2; Jdp-2; Jun dimerization protein 2; Jundm2; Jundp2; progesterone receptor co-activator; TIF |
| Gene Symbols | JDP2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.