Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JDP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563740
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563740 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563740 Supplier Invitrogen™ Supplier No. PA563740
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HRPTCIVRTD SVKTPESEGN PLLE Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

Component of the AP-1 transcription factor that represses transactivation mediated by the Jun family of proteins. Involved in a variety of transcriptional responses associated with AP-1 such as UV-induced apoptosis, cell differentiation, tumorigenesis and antitumogeneris. Can also function as a repressor by recruiting histone deacetylase 3/HDAC3 to the promoter region of JUN. May control transcription via direct regulation of the modification of histones and the assembly of chromatin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JDP2
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene JDP2
Gene Accession No. Q8WYK2
Gene Alias c-Jun dimerization protein 2; Jdp2; Jdp-2; Jun dimerization protein 2; Jundm2; Jundp2; progesterone receptor co-activator; TIF
Gene Symbols JDP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human JDP2. Recombinant protein control fragment (Product #RP-101814).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122953
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.