Learn More
Description
Application: Suitable for confocal microscopy, super-resolution microscopy (SRM) techniques including dSTORM (in both live and fixed cells) and STED. Cell permeable.
Properties and Photophysical Data: JFX™ 554, Maleimide shows enhanced brightness, photostability and chromostability. Maleimide can be converted to relevant substrate for use in self-labeling tag systems, e.g. HaloTag™ and SNAP-tag™. Excitation and emission maxima (λ) are 554 nm and 576 nm, respectively; quantum yield = 0.8; extinction coefficient = 104,600 M-1cm-1; lactone-zwitterion equilibrium constant (KL-Z) = 4.97.
Please see the product protocol for further information and a guide to protein/antibody labeling.
We also offer Janelia Fluor™ conjugated antibodies and custom conjugation services .
HaloTag is a trademark of Promega Corporation, and SNAP-tag is a trademark of New England BioLabs, Inc.
Janelia Fluor is a registered trademark of Howard Hughes Medical Institute.
JFX is a trademark owned by the Howard Hughes Medical Institute.
Specifications
Specifications
| Quantity | 200 μg |
| Product Line | Tocris |
| Target | Fluorescent Dyes |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
