Learn More
Tocris Bioscience™ JFX™ 650, NHS ester
Description
Application: Suitable for confocal microscopy, super resolution microscopy (SRM) techniques including dSTORM (in both live and fixed cells) and STED. Cell permeable.
Properties and Photophysical Data: JFX™ 650, NHS ester shows enhanced brightness, photostability and chromostability. NHS ester can be converted to relevant substrate for use in self-labeling tag systems, e.g. HaloTag™ and SNAP-tag™. Excitation and emission maxima (λ) are 650 nm and 667 nm, respectively; quantum yield = 0.53; extinction coefficient = 17,600 M-1cm-1; lactone-zwitterion equilibrium constant (KL-Z) = 0.015.
Please see the protocol for further information on protein/antibody labeling and conjugation.
We also offer Janelia Fluor™ conjugated antibodies and custom conjugation services with our sister company Novus Biologicals Biologicals.
HaloTag is a trademark of Promega Corporation, and SNAP-tag is a trademark of New England BioLabs, Inc.
Janelia Fluor is a registered trademark of Howard Hughes Medical Institute.
JFX is a trademark owned by the Howard Hughes Medical Institute.
Specifications
Specifications
| Chemical Name or Material | 2-(5,5-Dimethyl-3-(pyrrolidin-1-ium-1-ylidene-d8)-7-(pyrrolidin-1-yl-d8)-3,5-dihydrodibenzo[b,e]silin-10-yl)-4-(((2,5-dioxopyrrolidin-1-yl)oxy)carbonyl)benzoate |
| CAS | 2396479-84-6 |
| Quantity | 200 μg |
| Target | Amine-Reactive Janelia Fluor Dyes |
| Molecular Formula | C35H19D16N3O6Si |
| Purity | 0.9 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.