Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ JH-II-127
GREENER_CHOICESDP

Catalog No. 628950 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-895-0 50 mg
62-891-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 62-895-0 Supplier Tocris Bioscience™ Supplier No. 6289/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective LRRK2 inhibitor

JH-II-127 is a potent and selective LRRK2 inhibitor (IC50 values are 2.2, 6.6 and 47.7 nM for G2019S-mutant, wild-type and A2016T-mutant LRRK2, respectively). Selective for LRRK2 over a panel of 138 kinases. Brain penetrant and orally bioavailable.

Specifications

Chemical Name or Material [4-[[5-Chloro-4-(methylamino)-7H-pyrrolo[2,3-d]pyrimidin-2-yl]amino]-3-methoxyphenyl]-4-morpholinyl-methanone
CAS 1700693-08-8
Assay Percent Range >98%
Quantity 50 mg
Molecular Formula C19H21ClN6O3
Formula Weight Observed MW: 416.87
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.