Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ JHDM1D Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552912
Description
Immunogen sequence: LETLKELRED GFQPQTYLVQ GVKALHTALK LWMKKELVSE HAFEIPDNVR PGHLIKELSK VIRAIEEENG KPVKSQGIPI VCPVSRSSNE A Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 92%.
JHDM1D histone demethylase that specifically demethylates dimethylated 'Lys-9' and 'Lys-27' (H3K9me2 and H3K27me2, respectively) of histone H3, thereby playing a central role in histone code. This specifically binds trimethylated 'Lys-4' of histone H3 (H3K4me3), affecting histone demethylase specificity: in presence of H3K4me3, it has no demethylase activity toward H3K9me2, while it has high activity toward H3K27me2. This demethylates H3K9me2 in absence of H3K4me3.
Specifications
JHDM1D | |
Polyclonal | |
Unconjugated | |
KDM7A | |
A630082K20Rik; BB041802; ENSMUSG00000073143; histone lysine demethylase JHDM1D; Jhdm1d; JmjC domain-containing histone demethylation protein 1D; jumonji C domain containing histone demethylase 1; jumonji C domain containing histone demethylase 1 homolog D; jumonji C domain-containing histone demethylase 1 homolog D; Kdm7; Kdm7a; Kiaa1718; lysine (K)-specific demethylase 7A; lysine demethylase 7A; lysine-specific demethylase 7; Lysine-specific demethylase 7A; mKIAA1718 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80853 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZMT4 | |
KDM7A | |
Recombinant protein corresponding to Human JHDM1D. Recombinant protein control fragment (Product #RP-89695). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction