Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JHDM1D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576360
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576360 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576360 Supplier Invitrogen™ Supplier No. PA576360
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

JHDM1D histone demethylase that specifically demethylates dimethylated 'Lys-9' and 'Lys-27' (H3K9me2 and H3K27me2, respectively) of histone H3, thereby playing a central role in histone code. This specifically binds trimethylated 'Lys-4' of histone H3 (H3K4me3), affecting histone demethylase specificity: in presence of H3K4me3, it has no demethylase activity toward H3K9me2, while it has high activity toward H3K27me2. This demethylates H3K9me2 in absence of H3K4me3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JHDM1D
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene KDM7A
Gene Accession No. Q3UWM4, Q6ZMT4
Gene Alias A630082K20Rik; BB041802; ENSMUSG00000073143; histone lysine demethylase JHDM1D; Jhdm1d; JmjC domain-containing histone demethylation protein 1D; jumonji C domain containing histone demethylase 1; jumonji C domain containing histone demethylase 1 homolog D; jumonji C domain-containing histone demethylase 1 homolog D; Kdm7; Kdm7a; Kiaa1718; lysine (K)-specific demethylase 7A; lysine demethylase 7A; lysine-specific demethylase 7; Lysine-specific demethylase 7A; mKIAA1718
Gene Symbols KDM7A
Host Species Rabbit
Immunogen Recombinant fusion protein of mouse KDM7A.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338523, 684297, 80853
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.