Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JHDM1D Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$182.00 - $464.00
Specifications
Antigen | JHDM1D |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
JHDM1D Polyclonal specifically detects JHDM1D in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
JHDM1D | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
80853 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
jmjC domain-containing histone demethylation protein 1D, jumonji C domain containing histone demethylase 1 homolog D (S. cerevisiae), KDM7A | |
Recombinant fusion protein containing a sequence corresponding to amino acids 417-735 of mouse KDM7A JHDM1D (NP_001028602.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title