Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JIK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518294
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518294 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518294 Supplier Invitrogen™ Supplier No. PA518294
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

TAOK3 (JIK) is a serine/threonine kinase that is a member of the GCK-like subfamily of the STE20 kinase family. JIK is ubiquitously expressed in human tissues, inhibits JNK and has been implicated in cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JIK
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene TAOK3
Gene Accession No. Q9H2K8
Gene Alias 2900006A08Rik; A130052D22; A430105I05Rik; Arg357; Axotomy-related gene 357 protein; CTCL-associated antigen HD-CL-09; Cutaneous T-cell lymphoma-associated antigen HD-CL-09; dendritic cell-derived protein kinase; DKFZp666H245; DPK; EC 2.7.11.1; FLJ31808; hKFC-A; JIK; JNK/SAPK-inhibitory kinase; Jun kinase-inhibitory kinase; KDS; Kinase from chicken homolog A; MAP3K18; Serine/threonine-protein kinase TAO3; STE20-like kinase; STE2-like kinase; tao; TAO kinase 3; Taok3; Thousand and one amino acid protein 3
Gene Symbols TAOK3
Host Species Goat
Immunogen Synthetic peptide sequence (CHKKDHVFIRDEAGHGD) corresponding to the internal amino acids of TAOK3 (aa 402-412).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51347
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.