Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JIK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116005
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116005 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116005 Supplier Invitrogen™ Supplier No. PA5116005
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TAOK3.

TAOK3 (JIK) is a serine/threonine kinase that is a member of the GCK-like subfamily of the STE20 kinase family. JIK is ubiquitously expressed in human tissues, inhibits JNK and has been implicated in cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JIK
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene TAOK3
Gene Accession No. Q53UA7, Q8BYC6, Q9H2K8
Gene Alias 2900006A08Rik; A130052D22; A430105I05Rik; Arg357; Axotomy-related gene 357 protein; CTCL-associated antigen HD-CL-09; Cutaneous T-cell lymphoma-associated antigen HD-CL-09; dendritic cell-derived protein kinase; DKFZp666H245; DPK; EC 2.7.11.1; FLJ31808; hKFC-A; JIK; JNK/SAPK-inhibitory kinase; Jun kinase-inhibitory kinase; KDS; Kinase from chicken homolog A; MAP3K18; Serine/threonine-protein kinase TAO3; STE20-like kinase; STE2-like kinase; tao; TAO kinase 3; Taok3; Thousand and one amino acid protein 3
Gene Symbols TAOK3
Host Species Rabbit
Immunogen A synthesized peptide derived from human TAOK3(Accession Q9H2K8), corresponding to amino acid residues Q534-K584.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 304530, 330177, 51347
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.