Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JIP2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983047
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3047 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3047 Supplier Bioss Supplier No. BS13647R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is closely related to MAPK8IP1/IB1/JIP-1, a scaffold protein that is involved in the c-Jun amino-terminal kinase signaling pathway. This protein is expressed in brain and pancreatic cells. It has been shown to interact with, and regulate the activity of MAPK8/JNK1, and MAP2K7/MKK7 kinases. This protein thus is thought to function as a regulator of signal transduction by protein kinase cascade in brain and pancreatic beta-cells. Alternatively spliced transcript variants encoding distinct isoforms have been reported for this gene.

Specifications

Antigen JIP2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MAPK8IP2
Gene Accession No. Q13387
Gene Alias 3230402N03Rik; AI847694; AW049958; BOS_6281; C-Jun-amino-terminal kinase-interacting protein 2; homologous to mouse JIP-1; IB 2; IB2; IB-2; Islet brain 2; islet-brain 2; Islet-brain-2; JIP 2; Jip2; JIP-2; JNK MAP kinase scaffold protein 2; JNK MAP kinase scaffold protein JIP2; JNK-interacting protein 2; LOC100353641; Mapk8ip2; mitogen activated protein kinase 8 interacting protein 2; mitogen-activated protein kinase 8 interacting protein 2; mitogen-activated protein kinase 8 interacting protein 2-like; Mitogen-activated protein kinase 8-interacting protein 2; PRKM8IPL; R75148; si:dkey-180h12.2; si:dkey-181m21.1
Gene Symbols MAPK8IP2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human JIP2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23542
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.