Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JLP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575646
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575646 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575646 Supplier Thermo Scientific Supplier No. PA575646
Add to Cart
Add to Cart

JLP Polyclonal Antibody

JLP Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen JLP
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene SPAG9
Gene Accession No. O60271, Q58A65
Gene Alias 3110018C07Rik; 4733401I23Rik; 4831406C20Rik; AW552012; Cancer/testis antigen 89; c-Jun NH2-terminal kinase-associated leucine zipper protein; C-Jun-amino-terminal kinase-interacting protein 4; CT89; HLC4; HLC6; HLC-6; HSS; Human lung cancer oncogene 6 protein; Jip4; JIP-4; JLP; JNK interacting protein; JNK/SAPK-associated protein; JNK/SAPK-associated protein 2; JNK-associated leucine-zipper protein; JNK-interacting leucine zipper; JNK-interacting protein 4; Jsap2; JSAP2a; Kiaa0516; lung cancer oncogene 4; Mapk8ip4; Max-binding protein; mitogen-activated protein kinase 8-interacting protein 4; PHET; PIG6; proliferation-inducing gene 6; Proliferation-inducing protein 6; Protein highly expressed in testis; SPAG9; sperm associated antigen 9; sperm associated antigen 9 variant 5; sperm surface protein; Sperm-associated antigen 9; Sperm-specific protein; Sunday driver 1; syd1
Gene Symbols SPAG9
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues in Human JLP.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360600, 70834, 9043
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.