Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JLP Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501557406
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-7406 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-7406 Supplier Bethyl Laboratories, Inc Supplier No. A300337AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

Extracellular signals are transduced into cells through mitogen-activated protein kinases. The structural organization of these kinases into specific signaling domains is facilitated by scaffolding proteins involved in closely tethering different kinases so that successive phosphorylation events can occur. The protein encoded by this gene is a scaffolding protein that brings together mitogen-activated protein kinases and their transcription factor targets for the activation of specific signaling pathways. This gene which is abundantly expressed in testicular haploid germ cells encodes a protein that is recognized by sperm-agglutinating antibodies and implicated in infertility. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen JLP
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene SPAG9
Gene Accession No. O60271, Q58A65
Gene Alias C-jun-amino-terminal kinase-interacting protein 4; JNK-interacting protein 4; JIP-4; JNK-associated leucine-zipper protein; JLP; Sperm-associated antigen 9; Mitogen-activated protein kinase 8-interacting protein 4; Human lung cancer protein 6; HLC-6; Proliferation-inducing protein 6; Sperm-specific protein; Sperm surface protein; Protein highly expressed in testis; PHET; Sunday driver 1; Cancer/testis antigen 89; CT89; SPAG9; HSS; KIAA0516; MAPK8IP4; SYD1
Gene Symbols Spag9
Host Species Rabbit
Immunogen Between 775 and 825
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 70834, 9043
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.