Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JM109-lysate Rabbit anti-Bacteria, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | JM109-lysate |
|---|---|
| Concentration | 0.7 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-6408
![]()
|
Proteintech Group Inc
105611IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
JM109 competent E. coli contains mutations in recA1 and endA1 genes. These mutations aid in minimizing recombination and ensuring plasmid stability. The F? factor is suitable for the growth of bacteriophages (such as M13) to obtain single stranded DNA.Specifications
| JM109-lysate | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Bacteria | |
| JM109 lysate | |
| IgG | |
| Protein A | |
| Antibody |
| 0.7 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| JM109-lysate Fusion Protein Ag0046 | |
| Primary | |
| -20°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title