Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ JM4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116833
Description
Positive Samples: U-87MG, A-549, Mouse lung, Mouse brain, Rat lung, Rat brain Immunogen sequence: MSEVRLPPLR ALDDFVLGSA RLAAPDPCDP QRWCHRVINN LLYYQTNYLL.
JM4 is a 19 kDa four-transmembrane protein binding to the CCR5 receptor. It plays a role in the regulation of intracellular protein transport. It is expressed in normal brain, small intestine, lung, spleen, and pancreas and has also been detected in tumors of breast, colon, lung, and ovary.
Specifications
| JM4 | |
| Polyclonal | |
| Unconjugated | |
| PRAF2 | |
| AA986329; C78013; DXImx39e; Jena-Muenchen 4; JM4; PRA1 domain family 2; PRA1 domain family member 2; PRA1 domain family, member 2; PRA1 family protein 2; Praf2; RGD1566287; Sfc20; Yip6a | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11230, 367743, 54637 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.21 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O60831, Q9JIG8 | |
| PRAF2 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-50 of human PRAF2 (NP_0091441). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction