Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ JMJD1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520803
Description
A suggested positive control is rat liver tissue lysate. PA5-20803 can be used with blocking peptide PEP-0917.
Histone demethylase that specifically demethylates 'Lys-9' of histone H3, thereby playing a central role in histone code. Demethylation of Lys residue generates formaldehyde and succinate. May have tumor suppressor activity.
Specifications
| JMJD1B | |
| Polyclonal | |
| Unconjugated | |
| KDM3B | |
| 5830462I21Rik; 5qNCA; C5orf7; JHDM2B; JmjC domain-containing histone demethylation protein 2B; JMJD1B; jumonji domain containing 1B; jumonji domain-containing protein 1B; KDM3B; KDM3B lysine (K)-specific demethylase 3B; KIAA1082; lysine (K)-specific demethylase 3B; lysine demethylase 3B; lysine-specific demethylase 3B; mKIAA1082; NET22; Nuclear protein 5qNCA | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 277250, 51780, 682469 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q6ZPY7, Q7LBC6 | |
| KDM3B | |
| A 20 amino acid peptide from near the center of human JMJD1B. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction