Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JMJD1B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985681
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5681 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5681 Supplier Bioss Supplier No. BS3824R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histone demethylase that specifically demethylates ′Lys-9′ of histone H3, thereby playing a central role in histone code. Demethylation of Lys residue generates formaldehyde and succinate. May have tumor suppressor activity.

Specifications

Antigen JMJD1B
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene KDM3B
Gene Accession No. Q7LBC6
Gene Alias 5830462I21Rik; 5qNCA; C5orf7; JHDM2B; JmjC domain-containing histone demethylation protein 2B; JMJD1B; jumonji domain containing 1B; Jumonji domain-containing protein 1B; KDM3B; KDM3B lysine (K)-specific demethylase 3B; KIAA1082; lysine (K)-specific demethylase 3B; lysine demethylase 3B; Lysine-specific demethylase 3B; mKIAA1082; NET22; Nuclear protein 5qNCA
Gene Symbols KDM3B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human JMJD1B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51780
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.