Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JMJD1B Recombinant Rabbit Monoclonal Antibody (PSH0-11)
GREENER_CHOICE

Catalog No. PIMA545972
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545972 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545972 Supplier Invitrogen™ Supplier No. MA545972
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Predicted band size: 191 kDa Positive Control: Hela cell lysate, NIH/3T3 cell lysate, Hela. Subcellular Location: Nucleus. Sequence Similarities: 92.96% Mouse; 90.14% Rat. Tissue Specificity: Ubiquitous. Highly expressed in placenta, skeletal muscle, kidney, heart and liver.

Histone demethylase that specifically demethylates 'Lys-9' of histone H3, thereby playing a central role in histone code. Demethylation of Lys residue generates formaldehyde and succinate. May have tumor suppressor activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JMJD1B
Applications ChIP Assay, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone PSH0-11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide
Gene KDM3B
Gene Accession No. Q6ZPY7, Q7LBC6
Gene Alias 5830462I21Rik; 5qNCA; C5orf7; JHDM2B; JmjC domain-containing histone demethylation protein 2B; JMJD1B; jumonji domain containing 1B; jumonji domain-containing protein 1B; KDM3B; KDM3B lysine (K)-specific demethylase 3B; KIAA1082; lysine (K)-specific demethylase 3B; lysine demethylase 3B; lysine-specific demethylase 3B; mKIAA1082; NET22; Nuclear protein 5qNCA
Gene Symbols KDM3B
Host Species Rabbit
Immunogen Recombinant protein within human KDM3B aa 251-500.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 277250, 51780
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.