Learn More
Invitrogen™ JMJD1C Polyclonal Antibody

Description
A suggested positive control is human liver tissue lysate. PA5-20804 can be used with blocking peptide PEP-0918.
Specifications
Specifications
| Antigen | JMJD1C |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | JMJD1C |
| Gene Accession No. | Q15652, Q69ZK6 |
| Gene Alias | 5430433L24Rik; D630035I23Rik; DKFZp761F0118; FLJ14374; Jhdm2c; JMJD1C; Jmjdic; jumonji domain containing 1C; jumonji domain-containing protein 1C; Kiaa1380; Pr2; Probable JmjC domain-containing histone demethylation protein 2C; RP11-10C13.2; thyroid hormone receptor interactor 8; thyroid receptor-interacting protein 8; TR-interacting protein 8; TRIP8; TRIP-8 |
| Gene Symbols | JMJD1C |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.