Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JMJD1C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563258
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563258 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563258 Supplier Invitrogen™ Supplier No. PA563258
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PNVSDSKHSI ANAKFLETAK KDSDQSWVSD VVKVDLTQSS VTNASSGNDH LNMEKEKYVS YISPLSAVSV MEDKLHKRSP PPETIKSKLN TSVDTHKIKS SPSPEVVKPK Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 77%.

The jumonji domain containing 1C protein (JMJD1C) was initially discovered in silico, and later suggested to be a candidate gene for autism. Like the related proteins JMJD1A and JMJD1B, JMJD1C is a histone H3K9 demethylase implicated in the nuclear hormone receptor-based transcriptional regulation. JMJD1C mRNA is highly expressed in undifferentiated embryonic stem (ES) cells as well as pancreatic islet, diffuse-type gastric cancer, and other tissues and tumors. The JMJD1C gene promoter contain bHLH-, AP-1-, and POU5F1-binding sites, and as preferential expression of POU5F1 has been reported in ES cells, pancreatic islet, and diffuse-type gastric cancer, it has been suggested that POU5F1-mediated expression of JMJD1C reactivates previously silenced genes in ES cells and diffuse-type gastric cancer. At least three isoforms of JMJD1C are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JMJD1C
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene JMJD1C
Gene Accession No. Q15652
Gene Alias 5430433L24Rik; D630035I23Rik; DKFZp761F0118; FLJ14374; Jhdm2c; JMJD1C; Jmjdic; jumonji domain containing 1C; jumonji domain-containing protein 1C; Kiaa1380; Pr2; Probable JmjC domain-containing histone demethylation protein 2C; RP11-10C13.2; thyroid hormone receptor interactor 8; thyroid receptor-interacting protein 8; TR-interacting protein 8; TRIP8; TRIP-8
Gene Symbols JMJD1C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human JMJD1C. Recombinant protein control fragment (Product #RP-93608).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221037
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.