Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JMJD2A Monoclonal Antibody (C.37.0)
GREENER_CHOICE

Catalog No. PIMA514782
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514782 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514782 Supplier Invitrogen™ Supplier No. MA514782
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

It is not recommended to aliquot this antibody. This antibody is not cross-reactive with other Jumonji C proteins, including JMJD2B, JMJD2C and JMJD2D.

JMJD2A is a member of the Jumonji domain 2 (JMJD2) family and a protein containing a JmjN domain, a JmjC domain, a JD2H domain, two TUDOR domains, and two PHD-type zinc fingers. This protein functions as a trimethylation-specific demethylase, converting specific trimethylated histone residues to the dimethylated form, and as a transcriptional repressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JMJD2A
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone C.37.0
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5
Gene KDM4A
Gene Accession No. O75164, Q8BW72
Gene Alias D4Ertd222e; JHDM3A; jmjC domain-containing histone demethylation protein 3A; JMJD2; JMJD2A; jumonji C domain-containing histone demethylase 3A; jumonji domain containing 2; jumonji domain containing 2A; jumonji domain-containing protein 2A; KDM4A; KIAA0677; lysine (K)-specific demethylase 4A; lysine demethylase 4A; lysine-specific demethylase 4A; mKIAA0677; TDRD14A; tudor domain containing 14A
Gene Symbols KDM4A
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the human JMJD2A protein.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 230674, 313539, 9682
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.