Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JMJD2A Monoclonal Antibody (N154/32), Active Motif™
SDP

Mouse Monoclonal Antibody

$156.65 - $620.58

Specifications

Antigen JMJD2A
Clone N154/32
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-2878
SDP
Encompass
View Documents
Active Motif
39816
10 μg
Each for $156.65
Only null left
Add to Cart
 
50-199-2877
SDP
Encompass
View Documents
Active Motif
39815
100 μg
Each for $620.58
Only null left
Add to Cart
 
Description

Description

JMJD2A is a member of the Jumonji domain 2 (JMJD2) family and a protein containing a JmjN domain, a JmjC domain, a JD2H domain, two TUDOR domains, and two PHD-type zinc fingers. This protein functions as a trimethylation-specific demethylase, converting specific trimethylated histone residues to the dimethylated form, and as a transcriptional repressor.
Specifications

Specifications

JMJD2A
1 mg/mL
Monoclonal
Liquid
RUO
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
D4Ertd222e; Jhdm3a; JmjC domain-containing histone demethylation protein 3A; JMJD2; JMJD2A; jumonji C domain-containing histone demethylase 3A; jumonji domain containing 2; jumonji domain containing 2A; Jumonji domain-containing protein 2A; KDM4A; Kiaa0677; lysine (K)-specific demethylase 4A; lysine demethylase 4A; Lysine-specific demethylase 4A; mKIAA0677; TDRD14A; tudor domain containing 14A
KDM4A
IgG1
Protein G
Antibody
N154/32
Western Blot
Unconjugated
Mouse
Human, Mouse
O75164, Q8BW72
230674, 9682
This JMJD2A antibody was raised against a recombinant protein corresponding to amino acids 586-674 of human JMJD2A.
Primary
-20°C, Avoid Freeze/Thaw Cycles
KDM4A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.