Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JMJD2A Mouse anti-Human, Active Motif™
SDP

Catalog No. 501993853
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3853 100 μg
50-199-3854 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3853 Supplier Active Motif Supplier No. 91143
Add to Cart
Add to Cart

Mouse Recombinant Monoclonal Antibody

JMJD2A is a member of the Jumonji domain 2 (JMJD2) family and a protein containing a JmjN domain, a JmjC domain, a JD2H domain, two TUDOR domains, and two PHD-type zinc fingers. This protein functions as a trimethylation-specific demethylase, converting specific trimethylated histone residues to the dimethylated form, and as a transcriptional repressor.

Specifications

Antigen JMJD2A
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 200mM HEPES, 50mM sodium acetate with 100mM NaCl, 30% glycerol and 0.035% sodium azide; pH 7.5
Gene KDM4A
Gene Accession No. O75164
Gene Alias D4Ertd222e; Jhdm3a; JmjC domain-containing histone demethylation protein 3A; JMJD2; JMJD2A; jumonji C domain-containing histone demethylase 3A; jumonji domain containing 2; jumonji domain containing 2A; Jumonji domain-containing protein 2A; KDM4A; Kiaa0677; lysine (K)-specific demethylase 4A; lysine demethylase 4A; Lysine-specific demethylase 4A; mKIAA0677; TDRD14A; tudor domain containing 14A
Gene Symbols KDM4A
Host Species Mouse
Immunogen This JMJD2A antibody was raised against a recombinant protein corresponding to amino acids 586-674 of human JMJD2A.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9682
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.