Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ JMJD2A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520806
Description
A suggested positive control is rat liver tissue lysate. PA5-20806 can be used with blocking peptide PEP-0920.
JMJD2A is a member of the Jumonji domain 2 (JMJD2) family and a protein containing a JmjN domain, a JmjC domain, a JD2H domain, two TUDOR domains, and two PHD-type zinc fingers. This protein functions as a trimethylation-specific demethylase, converting specific trimethylated histone residues to the dimethylated form, and as a transcriptional repressor.
Specifications
JMJD2A | |
Polyclonal | |
Unconjugated | |
KDM4A | |
D4Ertd222e; JHDM3A; jmjC domain-containing histone demethylation protein 3A; JMJD2; JMJD2A; jumonji C domain-containing histone demethylase 3A; jumonji domain containing 2; jumonji domain containing 2A; jumonji domain-containing protein 2A; KDM4A; KIAA0677; lysine (K)-specific demethylase 4A; lysine demethylase 4A; lysine-specific demethylase 4A; mKIAA0677; TDRD14A; tudor domain containing 14A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
230674, 313539, 9682 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O75164, Q8BW72 | |
KDM4A | |
A 14 amino acid peptide from near the center of human JMJD2A. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction