Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JMJD2A Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA585519

Catalog No. PIPA585519


Only null left
Add to Cart

Description

Description

Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%), Bovine (100%). Positive Control: HeLa nuclear extract, Rat brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

JMJD2A is a member of the Jumonji domain 2 (JMJD2) family and a protein containing a JmjN domain, a JmjC domain, a JD2H domain, two TUDOR domains, and two PHD-type zinc fingers. This protein functions as a trimethylation-specific demethylase, converting specific trimethylated histone residues to the dimethylated form, and as a transcriptional repressor.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

JMJD2A
Polyclonal
Unconjugated
KDM4A
D4Ertd222e; JHDM3A; jmjC domain-containing histone demethylation protein 3A; JMJD2; JMJD2A; jumonji C domain-containing histone demethylase 3A; jumonji domain containing 2; jumonji domain containing 2A; jumonji domain-containing protein 2A; KDM4A; KIAA0677; lysine (K)-specific demethylase 4A; lysine demethylase 4A; lysine-specific demethylase 4A; mKIAA0677; TDRD14A; tudor domain containing 14A
Rabbit
Antigen affinity chromatography
RUO
313539, 9682
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
2.84 mg/mL
PBS with 20% glycerol and 0.01% thimerosal; pH 7
O75164
KDM4A
Recombinant protein encompassing a sequence within the C-terminus region of human KDM4A. The exact sequence is proprietary.
100 μL
Primary
Human, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.