Learn More
JMJD2A Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300861A
Description
The recommended shelf life for this product is 1 year from date of receipt.
KDM4A (Lysine-specific demethylase 4A) is a member of the Jumonji domain 2 family and encodes a protein containing a JmjN domain, a JmjC domain, a JD2H domain, two TUDOR domains, and two PHD-type zinc fingers. This nuclear protein functions as a trimethylation-specific demethylase, converting specific trimethylated histone residues to the dimethylated form. It also functions as a transcriptional repressor.Specifications
| JMJD2A | |
| Polyclonal | |
| Unconjugated | |
| KDM4A | |
| JMJD2A; JHDM3A, JMJD2, KIAA0677 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 230674, 9682 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| O75164, Q8BW72 | |
| Kdm4a | |
| Between 1010 and C-term | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.