Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JMJD2B Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10074605DL594
Description
JMJD2B Polyclonal antibody specifically detects JMJD2B in Human, Mouse samples. It is validated for Western BlotSpecifications
JMJD2B | |
Polyclonal | |
Western Blot | |
EC 1.14.11, FLJ44906, JHDM3B, JmjC domain-containing histone demethylation protein 3B, JMJD2BEC 1.14.11.-, jumonji domain containing 2B, Jumonji domain-containing protein 2B, KIAA0876lysine-specific demethylase 4B, lysine (K)-specific demethylase 4B | |
The immunogen recognized by this antibody maps to a region between residue 1056 and 1096 of human jumonji domain containing 2B using the numbering given in entry NP_055830.1 (GeneID 23030). | |
0.1 mL | |
Chromatin Research, Epigenetics | |
23030 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction