Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JMJD2B Antibody (SR1194), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | JMJD2B |
---|---|
Clone | SR1194 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
JMJD2B Monoclonal antibody specifically detects JMJD2B in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
JMJD2B | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 1.14.11, FLJ44906, JHDM3B, JmjC domain-containing histone demethylation protein 3B, JMJD2BEC 1.14.11.-, jumonji domain containing 2B, Jumonji domain-containing protein 2B, KIAA0876lysine-specific demethylase 4B, lysine (K)-specific demethylase 4B | |
A synthesized peptide derived from human JMJD2B (Uniprot #: O94953) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1194 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Chromatin Research, Epigenetics | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
23030 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title