Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JMJD2B / KDM4B Recombinant Mouse Monoclonal Antibody, AbFlex™, Active Motif™

Mouse Recombinant Monoclonal Antibody
$108.45 - $482.00
Specifications
Antigen | JMJD2B / KDM4B |
---|---|
Concentration | 1 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-2615
![]() ![]() |
Active Motif
91306 |
10 μg |
Each for $108.45
|
|
|||||
50-199-3811
![]() ![]() |
Active Motif
91305 |
100 μg |
Each for $482.00
|
|
|||||
Description
SOGA1 regulates autophagy by playing a role in the reduction of glucose production in an adiponectin- and insulin-dependent manner.Specifications
JMJD2B / KDM4B | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
O94953 | |
23030 | |
This antibody was raised against full-length recombinant human lysine-specific demethylase 4B (kDM4B) protein. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
KDM4B |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
140mM HEPES, 32mM sodium acetate with 30% glycerol, 70mM NaCl and 0.035% sodium azide; pH 7.5 | |
4732474L06Rik; JHDM3B; JmjC domain-containing histone demethylation protein 3B; Jmjd2b; jumonji domain containing 2B; Jumonji domain-containing protein 2B; KDM4B; KIAA0876; lysine (K)-specific demethylase 4B; lysine demethylase 4B; Lysine-specific demethylase 4B; mKIAA0876; TDRD14B; tudor domain containing 14B | |
KDM4B | |
IgG2a | |
Protein A | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title