Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JMJD2B Polyclonal Antibody
JMJD2B Polyclonal Antibody
Supplier: Thermo Scientific PA5115460
Description
JMJD2B Polyclonal Antibody for Western Blot, ICC/IF, IHC (PFA)

Specifications
JMJD2B | |
Polyclonal | |
Unconjugated | |
KDM4B | |
4732474L06Rik; JHDM3B; JmjC domain-containing histone demethylation protein 3B; Jmjd2b; jumonji domain containing 2B; jumonji domain-containing protein 2B; KDM4B; KIAA0876; lysine (K)-specific demethylase 4B; lysine demethylase 4B; lysine-specific demethylase 4B; mKIAA0876; TDRD14B; tudor domain containing 14B | |
Rabbit | |
Affinity Chromatography | |
RUO | |
193796, 23030, 301128 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
O94953, Q91VY5 | |
KDM4B | |
A synthesized peptide derived from human KDM4B(Accession O94953), corresponding to amino acid residues M585-E635. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction