Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JMJD2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120953
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120953 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120953 Supplier Invitrogen™ Supplier No. PA5120953
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse brain. The target is usually found in the following locations: Nucleus. Immunogen sequence: MGSEDHGAQN PSCKIMTFRP TMEEFKDFNK YVAYIESQGA HRAGLAKIIP PKEWKPRQTY DDIDDVVIPA PIQQVVTGQS GLFTQYNIQK KAMTVGEYRR LANSEKYCTP RHQDFDDLER KYWKNLTFVS PIYGADISGS LYDDDVAQWN IGSLRTILDM VERECGTIIE GVNTPYLYFG MWKTTFAWHT EDMDLYSINY LHFGEPKSWY AIPPEHGKRL ERLAIGFFPG SSQGCDAFLR HKMTLISPII LKKYGIPFSR ITQEAGEFMI TFPYGYHAGF NHGFNCAEST NFATLRWIDY GKVATQCTCR KDMVKISMDV FVRILQPERY ELWKQGKDLT VLDHTRPTAL TSPELSSWSA SRASL.

Histone demethylase that specifically demethylates 'Lys-9' of histone H3, thereby playing a role in histone code. Does not demethylate histone H3 'Lys-4', H3 'Lys-27', H3 'Lys-36' nor H4 'Lys-20'. Only able to demethylate trimethylated H3 'Lys-9', with a weaker activity than KDM4A, KDM4C and KDM4D. Demethylation of Lys residue generates formaldehyde and succinate. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen JMJD2B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene KDM4B
Gene Accession No. O94953, Q91VY5
Gene Alias 4732474L06Rik; JHDM3B; JmjC domain-containing histone demethylation protein 3B; Jmjd2b; jumonji domain containing 2B; jumonji domain-containing protein 2B; KDM4B; KIAA0876; lysine (K)-specific demethylase 4B; lysine demethylase 4B; lysine-specific demethylase 4B; mKIAA0876; TDRD14B; tudor domain containing 14B
Gene Symbols KDM4B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-365 of human KDM4B/JMJD2B (NP_0558301).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 193796, 23030
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.