Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JMJD2C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567044
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567044 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567044 Supplier Invitrogen™ Supplier No. PA567044
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ARFSTASDMRF EDTFYGADII QGERKRQRVL SSRFKNEYVA DPVYRTFLKS SFQKKCQK Highest antigen sequence identity to the following orthologs - mouse 95%, rat 95%.

Jumonji domain-containing protein 2C (JMJD2C)/ Lysine-specific demethylase 4C (KDM4C) is a histone demethylase that specifically demethylates 'Lys-9' and 'Lys-36' residues of histone H3; thereby, playing a central role in histone code. KDM4C does not demethylate histone H3 'Lys-4', H3 'Lys-27', or H4 'Lys-20'. It demethylates trimethylated H3 'Lys-9' and H3 'Lys-36' residues. There is no activity on mono- or dimethylated residues. Demethylation of Lys residues generates formaldehyde and succinate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JMJD2C
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KDM4C
Gene Accession No. Q9H3R0
Gene Alias 2410141F18Rik; AA517467; bA146B14.1; GASC1; GASC-1 protein; gene amplified in squamous cell carcinoma 1; gene amplified in squamous cell carcinoma 1 protein; JHDM3C; JmjC domain-containing histone demethylation protein 3C; JMJD2C; jumonji domain containing 2C; jumonji domain-containing protein 2C; Kdm4c; KIAA0780; lysine (K)-specific demethylase 4C; lysine demethylase 4C; lysine-specific demethylase 4C; RP11-169L18.1; TDRD14C; tudor domain containing 14C
Gene Symbols KDM4C
Host Species Rabbit
Immunogen Recombinant Human JMJD2C. Recombinant protein control fragment (Product #RP-107688).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23081
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.