Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JMJD2C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115461
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115461 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115461 Supplier Invitrogen™ Supplier No. PA5115461
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KDM4C / GASC1 / JMJD2C.

Jumonji domain-containing protein 2C (JMJD2C)/ Lysine-specific demethylase 4C (KDM4C) is a histone demethylase that specifically demethylates 'Lys-9' and 'Lys-36' residues of histone H3; thereby, playing a central role in histone code. KDM4C does not demethylate histone H3 'Lys-4', H3 'Lys-27', or H4 'Lys-20'. It demethylates trimethylated H3 'Lys-9' and H3 'Lys-36' residues. There is no activity on mono- or dimethylated residues. Demethylation of Lys residues generates formaldehyde and succinate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JMJD2C
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene KDM4C
Gene Accession No. Q8VCD7, Q9H3R0
Gene Alias 2410141F18Rik; AA517467; bA146B14.1; GASC1; GASC-1 protein; gene amplified in squamous cell carcinoma 1; gene amplified in squamous cell carcinoma 1 protein; JHDM3C; JmjC domain-containing histone demethylation protein 3C; JMJD2C; jumonji domain containing 2C; jumonji domain-containing protein 2C; Kdm4c; KIAA0780; lysine (K)-specific demethylase 4C; lysine demethylase 4C; lysine-specific demethylase 4C; RP11-169L18.1; TDRD14C; tudor domain containing 14C
Gene Symbols KDM4C
Host Species Rabbit
Immunogen A synthesized peptide derived from human KDM4C(Accession Q9H3R0), corresponding to amino acid residues Y472-G522.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23081, 298144, 76804
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.