Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JMJD2C Recombinant Rabbit Monoclonal Antibody (23H4L4)

Catalog No. 703380
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-338-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-338-0 Supplier Invitrogen™ Supplier No. 703380
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Monkey, Pig, Cat, Sheep. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Jumonji domain-containing protein 2C (JMJD2C)/ Lysine-specific demethylase 4C (KDM4C) is a histone demethylase that specifically demethylates 'Lys-9' and 'Lys-36' residues of histone H3; thereby, playing a central role in histone code. KDM4C does not demethylate histone H3 'Lys-4', H3 'Lys-27', or H4 'Lys-20'. It demethylates trimethylated H3 'Lys-9' and H3 'Lys-36' residues. There is no activity on mono- or dimethylated residues. Demethylation of Lys residues generates formaldehyde and succinate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JMJD2C
Applications Western Blot
Classification Recombinant Monoclonal
Clone 23H4L4
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene KDM4C
Gene Accession No. Q9H3R0
Gene Alias 2410141F18Rik; AA517467; bA146B14.1; GASC1; GASC-1 protein; gene amplified in squamous cell carcinoma 1; gene amplified in squamous cell carcinoma 1 protein; JHDM3C; JmjC domain-containing histone demethylation protein 3C; JMJD2C; jumonji domain containing 2C; jumonji domain-containing protein 2C; Kdm4c; KIAA0780; lysine (K)-specific demethylase 4C; lysine demethylase 4C; lysine-specific demethylase 4C; RP11-169L18.1; TDRD14C; tudor domain containing 14C
Gene Symbols KDM4C
Host Species Rabbit
Immunogen Peptides corresponding to human KDM4C [1) aa1019-aa1038 2) aa494-aa513].
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23081
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.